Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asgr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246579.1 | XP_006246641.1 | 1 - 2 | 158 | 127 | |
GenBank mRNA | FQ209763.1 | - | 1 - 2 | 82 | 127 | |
FQ218553.1 | - | 1 - 2 | 122 | 127 | ||
FQ218644.1 | - | 1 - 2 | 82 | 127 | ||
FQ218824.1 | - | 1 - 2 | 82 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |