Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsen54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109576.1 | NP_001103046.1 | 2 - 3 | 278 | 73 | |
XM_006247737.2 | XP_006247799.1 | 2 - 3 | 1201 | 73 | ||
GenBank mRNA | BC166577.1 | - | 2 - 3 | 222 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |