Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105856.1 | NP_001099326.1 | 1 - 2 | 103 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |