Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlst8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022404.2 | NP_071799.2 | 3 - 4 | 545 | 64 | |
GenBank mRNA | BC097319.1 | - | 3 - 4 | 223 | 7455095 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |