Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pisd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001061408.5 | XP_001061408.2 | 5 - 6 | 724 | 139 | |
XM_006251423.2 | XP_006251485.1 | 5 - 6 | 4654 | 139 | ||
XM_006251424.2 | XP_006251486.1 | 5 - 6 | 3836 | 139 | ||
XM_006251426.2 | XP_006251488.1 | 4 - 5 | 4418 | 139 | ||
XM_008770389.1 | XP_008768611.1 | 4 - 5 | 3963 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |