Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uspl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105906.2 | NP_001099376.1 | 6 - 7 | 1273 | 131 | |
NM_001198555.1 | NP_001185484.1 | 6 - 7 | 1273 | 131 | ||
XM_006248810.2 | XP_006248872.1 | 5 - 6 | 1216 | 131 | ||
GenBank mRNA | CO396482.1 | - | 2 - 3 | 186 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |