Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008347.1 | NP_001008348.1 | 6 - 7 | 609 | 107 | |
XM_006230439.2 | XP_006230501.1 | 6 - 7 | 654 | 107 | ||
GenBank mRNA | BC082095.1 | - | 6 - 7 | 609 | 7134234 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |