Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106372.2 | NP_001099842.1 | 4 - 5 | 836 | 103 | |
XM_008760367.1 | XP_008758589.1 | 4 - 5 | 887 | 103 | ||
GenBank mRNA | BC158801.1 | - | 4 - 5 | 836 | 7321043 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |