Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asrgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145089.4 | NP_659557.1 | 3 - 4 | 449 | 104 | |
GenBank mRNA | AF329099.1 | - | 3 - 4 | 449 | 104 | |
AJ427914.2 | - | 463 | 104 | |||
FQ212762.1 | - | 3 - 4 | 423 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |