Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053509.1 | NP_445961.1 | 7 - 8 | 1244 | 110 | |
GenBank mRNA | AB000928.1 | - | 7 - 8 | 1244 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |