Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031008.2 | NP_112270.2 | 16 - 17 | 2287 | 89 | |
XM_008760010.1 | XP_008758232.1 | 16 - 17 | 2342 | 89 | ||
GenBank mRNA | BC081786.1 | - | 16 - 17 | 2287 | 7126468 | 89 |
X53773.1 | - | 2240 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |