Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011976.2 | NP_001011976.2 | 6 - 7 | 660 | 112 | |
XM_006238237.2 | XP_006238299.1 | 6 - 7 | 624 | 112 | ||
XM_006238239.2 | XP_006238301.1 | 6 - 7 | 686 | 112 | ||
GenBank mRNA | BC079286.1 | - | 6 - 7 | 660 | 7134286 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |