Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bnc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106666.1 | NP_001100136.1 | 3 - 4 | 418 | 76 | |
XM_008763776.1 | XP_008761998.1 | 2 - 3 | 1609 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |