Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtcap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106444.1 | NP_001099914.1 | 3 - 4 | 734 | 106 | |
GenBank mRNA | BC167108.1 | - | 3 - 4 | 295 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |