Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191708.1 | NP_001178637.1 | 4 - 5 | 475 | 71 | |
XM_008761143.1 | XP_008759365.1 | 5 - 6 | 569 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus 1700 Probe spans exons |