Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gon4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024797.1 | NP_001019968.1 | 25 - 26 | 5440 | 95 | |
XM_006232735.2 | XP_006232797.1 | 25 - 26 | 5431 | 95 | ||
XM_006232736.2 | XP_006232798.1 | 25 - 26 | 5416 | 95 | ||
XM_006232737.2 | XP_006232799.1 | 24 - 25 | 5395 | 95 | ||
XM_006232738.2 | XP_006232800.1 | 24 - 25 | 5392 | 95 | ||
XM_006232739.2 | XP_006232801.1 | 25 - 26 | 5371 | 95 | ||
XM_006232740.2 | XP_006232802.1 | 24 - 25 | 5227 | 95 | ||
XM_008761201.1 | XP_008759423.1 | 26 - 27 | 5445 | 95 | ||
XM_008761202.1 | XP_008759424.1 | 24 - 25 | 4880 | 95 | ||
GenBank mRNA | AB195689.1 | - | 24 - 25 | 5274 | 95 | |
AY724781.1 | - | 25 - 26 | 5440 | 95 | ||
AY724782.1 | - | 25 - 26 | 5440 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |