Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024747.1 | NP_001019918.1 | 6 - 7 | 851 | 65 | |
GenBank mRNA | BC098049.1 | - | 6 - 7 | 851 | 7457308 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |