Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168642.1 | NP_001162113.1 | 2 - 3 | 276 | 128 | |
XM_006228106.2 | XP_006228168.1 | 2 - 3 | 322 | 128 | ||
XM_008758775.1 | XP_008756997.1 | 2 - 3 | 323 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |