Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134553.1 | NP_001128025.1 | 7 - 8 | 1666 | 116 | |
XM_006236308.2 | XP_006236370.1 | 7 - 8 | 2749 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |