Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004233.1 | NP_001004233.1 | 1 - 2 | 188 | 58 | |
XM_006236301.2 | XP_006236363.1 | 1 - 2 | 388 | 58 | ||
GenBank mRNA | BC079407.1 | - | 1 - 2 | 188 | 7112867 | 58 |
FQ226725.1 | - | 1 - 2 | 339 | 58 | ||
FQ233779.1 | - | 1 - 2 | 337 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |