Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pon2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013082.1 | NP_001013100.1 | 2 - 3 | 192 | 143 | |
XM_008762699.1 | XP_008760921.1 | 2 - 3 | 305 | 143 | ||
GenBank mRNA | BC079462.1 | - | 2 - 3 | 192 | 7122087 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |