Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017062.2 | NP_058758.1 | 6 - 7 | 855 | 114 | |
XM_006246230.2 | XP_006246292.1 | 6 - 7 | 1200 | 114 | ||
XM_006246231.2 | XP_006246293.1 | 6 - 7 | 913 | 114 | ||
GenBank mRNA | X76454.1 | - | 6 - 7 | 856 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |