Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768238.1 | XP_008766460.1 | 6 - 7 | 2456 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |