Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpain | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033060.1 | NP_001028232.1 | 5 - 6 | 489 | 119 | |
XM_006246668.2 | XP_006246730.1 | 5 - 6 | 853 | 119 | ||
GenBank mRNA | AY775322.1 | - | 5 - 6 | 489 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |