Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spns3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109035.1 | NP_001102505.1 | 6 - 7 | 965 | 72 | |
XM_006246786.2 | XP_006246848.1 | 7 - 8 | 1554 | 72 | ||
XM_008767857.1 | XP_008766079.1 | 7 - 8 | 1555 | 72 | ||
XM_008767858.1 | XP_008766080.1 | 7 - 8 | 1555 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Transporter 3' Most Rattus norvegicus Probe spans exons |