Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnptg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100493.1 | NP_001093963.1 | 2 - 3 | 128 | 97 | |
XM_006245935.2 | XP_006245997.1 | 2 - 3 | 145 | 97 | ||
GenBank mRNA | BC098716.1 | - | 2 - 3 | 110 | 97 | |
BC158818.1 | - | 2 - 3 | 128 | 7374209 | 97 | |
FQ213320.1 | - | 2 - 3 | 135 | 97 | ||
FQ213586.1 | - | 2 - 3 | 93 | 97 | ||
FQ213702.1 | - | 2 - 3 | 127 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |