Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271027.1 | NP_001257956.1 | 2 - 3 | 669 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Rattus norvegicus Probe spans exons |