Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107128.2 | NP_001100598.2 | 3 - 4 | 289 | 75 | |
XM_008768975.1 | XP_008767197.1 | 3 - 4 | 347 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |