Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micall2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248975.2 | XP_006249037.1 | 7 - 8 | 2208 | 77 | |
GenBank mRNA | FQ232177.1 | - | 1 - 2 | 35 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |