Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037217.1 | NP_001032294.1 | 1 - 2 | 217 | 69 | |
GenBank mRNA | BC161816.1 | - | 1 - 2 | 195 | 7320318 | 69 |
CB748602.1 | - | 1 - 2 | 275 | 69 | ||
CO400908.1 | - | 1 - 2 | 194 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |