Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133393.2 | NP_596884.1 | 4 - 5 | 786 | 59 | |
GenBank mRNA | AB054539.1 | - | 4 - 5 | 727 | 59 | |
BC070933.1 | - | 4 - 5 | 786 | 7097855 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |