Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021677.1 | NP_067709.1 | 2 - 3 | 190 | 62 | |
GenBank mRNA | U66292.1 | - | 2 - 3 | 190 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |