Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106231.2 | NP_001099701.2 | 1172 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Phosphatase Detect Genomic DNA Rattus norvegicus Probe spans exons |