Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001053214.4 | XP_001053214.2 | 8 - 9 | 1183 | 94 | |
XM_006228482.1 | XP_006228544.1 | 8 - 9 | 1355 | 94 | ||
XM_006228483.2 | XP_006228545.1 | 9 - 10 | 1892 | 94 | ||
XM_006228484.2 | XP_006228546.1 | 9 - 10 | 1246 | 94 | ||
XM_006228485.2 | XP_006228547.1 | 9 - 10 | 1575 | 94 | ||
XM_006228486.2 | XP_006228548.1 | 8 - 9 | 1747 | 94 | ||
XM_006228487.1 | XP_006228549.1 | 8 - 9 | 1106 | 94 | ||
XM_006228488.2 | XP_006228550.1 | 9 - 10 | 1458 | 94 | ||
XM_006228489.1 | XP_006228551.1 | 9 - 10 | 1245 | 94 | ||
XM_006228491.2 | XP_006228553.1 | 9 - 10 | 1369 | 94 | ||
XM_006228493.1 | XP_006228555.1 | 8 - 9 | 1355 | 94 | ||
XM_008759031.1 | XP_008757253.1 | 8 - 9 | 1300 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |