Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012459.1 | NP_001012477.1 | 3 - 4 | 659 | 124 | |
GenBank mRNA | AB009246.1 | - | 3 - 4 | 541 | 124 | |
BC090344.1 | - | 3 - 4 | 659 | 7304238 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |