Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ffar3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108912.1 | NP_001102382.1 | 1 - 2 | 49 | 62 | |
GenBank mRNA | BC166522.1 | - | 1 - 2 | 170 | 7134236 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |