Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spred3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173344.1 | NP_001166815.1 | 4 - 5 | 1002 | 104 | |
XM_008759144.1 | XP_008757366.1 | 5 - 6 | 640 | 104 | ||
XM_008759145.1 | XP_008757367.1 | 5 - 6 | 587 | 104 | ||
XM_008759146.1 | XP_008757368.1 | 5 - 6 | 709 | 104 | ||
GenBank mRNA | BC166515.1 | - | 4 - 5 | 1002 | 7131550 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |