Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109422.1 | NP_001102892.1 | 3 - 4 | 545 | 96 | |
XM_006228473.2 | XP_006228535.1 | 3 - 4 | 638 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Rattus norvegicus Probe spans exons |