Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptov1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008304.1 | NP_001008305.1 | 7 - 8 | 911 | 65 | |
GenBank mRNA | BC085836.1 | - | 7 - 8 | 911 | 7135127 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |