Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728716.4 | XP_002728762.3 | 2 - 3 | 2136 | 96 | |
XM_006228522.2 | XP_006228584.1 | 2 - 3 | 2136 | 96 | ||
XM_006228525.1 | XP_006228587.1 | 3 - 4 | 427 | 96 | ||
XM_006228526.1 | XP_006228588.1 | 3 - 4 | 417 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus Probe spans exons |