Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nccrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134506.2 | NP_001127978.1 | 448 | 113 | ||
XM_008759218.1 | XP_008757440.1 | 2 - 3 | 448 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |