Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spint2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082549.1 | NP_001076018.1 | 1 - 2 | 391 | 64 | |
GenBank mRNA | DN934634.1 | - | 1 - 2 | 420 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |