Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spint2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082549.1 | NP_001076018.1 | 3 - 4 | 617 | 57 | |
NM_199087.1 | NP_954518.1 | 2 - 3 | 446 | 57 | ||
GenBank mRNA | BC061768.1 | - | 2 - 3 | 446 | 5621353 | 57 |
DN934634.1 | - | 3 - 4 | 646 | 57 | ||
FQ229514.1 | - | 3 - 4 | 345 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |