Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014159.2 | NP_001014181.2 | 1 - 2 | 231 | 92 | |
XM_006228781.2 | XP_006228843.1 | 1 - 2 | 256 | 92 | ||
GenBank mRNA | BC079321.1 | - | 1 - 2 | 189 | 7135399 | 92 |
FQ222852.1 | - | 1 - 2 | 201 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |