Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014159.2 | NP_001014181.2 | 9 - 10 | 1421 | 100 | |
XM_006228781.2 | XP_006228843.1 | 8 - 9 | 1359 | 100 | ||
GenBank mRNA | BC079321.1 | - | 9 - 10 | 1379 | 7135399 | 100 |
BI273871.1 | - | 1 - 2 | 100 | |||
FQ216075.1 | - | 5 - 6 | 803 | 100 | ||
FQ235070.1 | - | 5 - 6 | 771 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |