Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031648.2 | NP_113836.2 | 4 - 5 | 257 | 68 | |
XM_006228838.2 | XP_006228900.1 | 5 - 6 | 477 | 68 | ||
XM_006228839.2 | XP_006228901.1 | 4 - 5 | 252 | 68 | ||
XM_008759184.1 | XP_008757406.1 | 4 - 5 | 266 | 68 | ||
GenBank mRNA | CB712570.1 | - | 4 - 5 | 257 | 68 | |
FM109156.1 | - | 4 - 5 | 211 | 68 | ||
FQ211349.1 | - | 4 - 5 | 268 | 68 | ||
FQ224131.1 | - | 4 - 5 | 220 | 68 | ||
FQ224338.1 | - | 4 - 5 | 303 | 68 | ||
U72246.1 | - | 3 - 4 | 171 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |