Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nosip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106260.1 | NP_001099730.1 | 3 - 4 | 225 | 80 | |
XM_006229025.1 | XP_006229087.1 | 3 - 4 | 378 | 80 | ||
XM_006229026.2 | XP_006229088.1 | 4 - 5 | 410 | 80 | ||
GenBank mRNA | FQ216351.1 | - | 3 - 4 | 345 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |