Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053929.1 | NP_446381.1 | 8 - 9 | 916 | 90 | |
XM_006228875.2 | XP_006228937.1 | 9 - 10 | 1018 | 90 | ||
XM_006228876.2 | XP_006228938.1 | 9 - 10 | 1398 | 90 | ||
XM_006228877.2 | XP_006228939.1 | 8 - 9 | 1277 | 90 | ||
XM_006228878.2 | XP_006228940.1 | 5 - 6 | 583 | 90 | ||
XM_008759103.1 | XP_008757325.1 | 8 - 9 | 1302 | 90 | ||
XM_008759104.1 | XP_008757326.1 | 8 - 9 | 957 | 90 | ||
GenBank mRNA | AB029559.1 | - | 8 - 9 | 916 | 90 | |
BC081750.1 | - | 7 - 8 | 1332 | 90 | ||
BC098909.1 | - | 8 - 9 | 906 | 7109606 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |