Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ambp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012901.1 | NP_037033.1 | 9 - 10 | 1042 | 94 | |
GenBank mRNA | BC088166.1 | - | 9 - 10 | 1027 | 7368246 | 94 |
FQ209788.1 | - | 6 - 7 | 610 | 94 | ||
FQ210037.1 | - | 6 - 7 | 610 | 94 | ||
FQ210364.1 | - | 6 - 7 | 610 | 94 | ||
FQ210824.1 | - | 6 - 7 | 610 | 94 | ||
FQ219366.1 | - | 6 - 7 | 610 | 94 | ||
FQ219628.1 | - | 6 - 7 | 611 | 94 | ||
FQ219767.1 | - | 6 - 7 | 610 | 94 | ||
S87544.1 | - | 9 - 10 | 1042 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |