Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134883.1 | NP_001128355.1 | 3 - 4 | 346 | 85 | |
XM_006241606.2 | XP_006241668.1 | 2 - 3 | 242 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Protease Rattus norvegicus Probe spans exons |